5-Amino-3-methylisoxazol ≥97.0% (by titrimetric analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
3-Methylisoxazol-5-ylamine, 3-Methylisoxazol-5-amine
A2867-1G
A2867-5G
TCIAA2867-1GEA
296
DKK
TCIAA2867-1G
TCIAA2867-5G
5-Amino-3-methylisoxazol ≥97.0% (by titrimetric analysis)
5-Amino-3-methylisoxazole
Formel:
C₄H₆N₂O |
MDL Number:
MFCD00003151 CAS-nummer: 14678-02-5 EINECS: 238-719-9 |
Specifikationer Testresultater
Purity (GC) | min. 95.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Melting point | 82.0 to 86.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...