2-Amino-4-chloro-5-formylthiophene-3-carbonitrile ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
2-Amino-4-chloro-5-formyl-3-thiophenecarbonitrile, 2-Amino-4-chloro-3-cyano-5-formylthiophene
A2863-1G
A2863-5G
TCIAA2863-1GEA
485
DKK
TCIAA2863-1G
TCIAA2863-5G
2-Amino-4-chloro-5-formylthiophene-3-carbonitrile ≥98.0% (by GC)
2-Amino-4-chloro-5-formylthiophene-3-carbonitrile
Formel:
C₆H₃ClN₂OS Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD03788265 CAS-nummer: 104366-23-6 UN nummer: 2923 ADR: 8,III |
Specifikationer Testresultater
Appearance | Light yellow to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...