2-Acetylphenothiazin ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Methyl phenothiazin-2-yl ketone, 1-(10H-Phenothiazin-2-yl)ethanone, ML171

A2815-5G A2815-25G
TCIAA2815-5GEA 1110 DKK
TCIAA2815-5G TCIAA2815-25G
2-Acetylphenothiazin ≥98.0% (by GC)
2-Acetylphenothiazin
Formel: C₁₄H₁₁NOS
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00005017
CAS-nummer: 6631-94-3
EINECS: 229-626-4

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 190.0 to 194.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR