1,3-Benzoxazol-2-amine ≥98.0% (by HPLC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Benzo[d]oxazol-2-amine, 2-Aminobenzoxazole
A2774-5G
A2774-25G
TCIAA2774-5GEA
1100
DKK
TCIAA2774-5G
TCIAA2774-25G
1,3-Benzoxazol-2-amine ≥98.0% (by HPLC)
1,3-Benzoxazol-2-amine
Formel:
C₇H₆N₂O Molekylvægt: 134.1353 g/mol |
MDL Number:
MFCD00182850 CAS-nummer: 4570-41-6 EINECS: 224-952-3 |
Specifikationer Testresultater
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 97.0 % |
Melting point | 128.0 to 132.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...