4-Amino-3-iodbenzonitril ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 4-Cyano-2-iodoaniline

A2747-1G A2747-5G
TCIAA2747-1GEA 386 DKK
TCIAA2747-1G TCIAA2747-5G
4-Amino-3-iodbenzonitril ≥98.0% (by GC)
4-Amino-3-iodbenzonitril
Formel: C₇H₅IN₂
Molekylvægt: 244.03 g/mol
MDL Number: MFCD04039964
CAS-nummer: 33348-34-4
UN nummer: 3439
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=18b1e12a2b743bd41328ccbf59f82daf79fe61fa&vid=18b1e12a2b743bd41328ccbf59f82daf79fe61fa&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Amber powder to crystal
Purity (GC) min. 98.0 %
Melting point 113.0 to 117.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR