3-Amino-4-(isobutylamino)quinoline hydrochloride ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
A2721-1G
A2721-5G
TCIAA2721-1GEA
181
DKK
TCIAA2721-1G
TCIAA2721-5G
3-Amino-4-(isobutylamino)quinoline hydrochloride ≥98.0% (by HPLC, titration analysis)
3-Amino-4-(isobutylamino)quinoline hydrochloride
Formel:
C₁₃H₁₇N₃ |
MDL Number:
MFCD08063180 CAS-nummer: 935521-01-0 |
Specifikationer Testresultater
Appearance | White to Amber to Dark green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...