tert-Butyl-6-aminopyridin-2-ylcarbamate 97,0 Titrimetric

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-Amino-6-(tert-butoxycarbonylamino)pyridine, 2-Amino-6-(Boc-amino)pyridine, tert-Butyl 6-aminopyridin-2-ylcarbamate

A2708-1G A2708-5G
TCIAA2708-1GEA 550 DKK
TCIAA2708-1G TCIAA2708-5G
tert-Butyl-6-aminopyridin-2-ylcarbamate 97,0 Titrimetric
tert-Butyl-6-aminopyridin-2-ylcarbamate
Formel: C₁₀H₁₅N₃O₂
Molekylvægt: 209.2500 g/mol
MDL Number: MFCD09037865
CAS-nummer: 322690-31-3

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 125.0 to 129.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR