N-Acetyltryptamine ≥98.0% (by HPLC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: N-(2-(1H-Indol-3-yl)ethyl)acetamide, 3-(2-Acetamidoethyl)indole

A2685-25MG A2685-100MG
TCIAA2685-25MGEA 493 DKK
TCIAA2685-25MG TCIAA2685-100MG
N-Acetyltryptamine ≥98.0% (by HPLC)
N-Acetyltryptamine
Formel: C₁₂H₁₄N₂O
Kogepunkt: 200°C
MDL Number: MFCD00209910
CAS-nummer: 1016-47-3

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 13.30 to 14.30 %
Melting point 74.0 to 78.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR