1,1'-Diethyl-3,3,3',3'-tetramethylindocarbocyanine iodide ≥98.0% (by HPLC, total nitrogen)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
DiIC2(3), 1-Ethyl-2-[3-(1-ethyl-1,3-dihydro-3,3-dimethyl-2H-indol-2-ylidene)-1-propen-1-yl]-3,3-dimethyl-3H-indolium iodide, Astrophloxine
A2684-1G
A2684-5G
TCIAA2684-1GEA
575
DKK
TCIAA2684-1G
TCIAA2684-5G
1,1'-Diethyl-3,3,3',3'-tetramethylindocarbocyanine iodide ≥98.0% (by HPLC, total nitrogen)
1,1'-Diethyl-3,3,3',3'-tetramethylindocarbocyanine iodide
Formel:
C₂₇H₃₃IN₂ |
MDL Number:
MFCD22573598 CAS-nummer: 14696-39-0 |
Specifikationer Testresultater
Appearance | Red to Dark red to Dark purple powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...