≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Phenylacetic acid Pentyl ester, Phenylacetic acid Amyl ester, Amyl Phenylacetate, Pentyl Phenylacetate

A2680-5ML A2680-25ML
TCIAA2680-5MLEA 312 DKK
TCIAA2680-5ML TCIAA2680-25ML
≥98.0% (by GC)
Pentylphenylacetat
Formel: C₁₃H₁₈O₂
Kogepunkt: 269°C
MDL Number: MFCD00027272
CAS-nummer: 5137-52-0
EINECS: 225-895-7

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR