N-Acetyl-3,5-dimethyl-1-adamantanamine ≥97.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: N-Acetylmemantine, 1-Acetamido-3,5-dimethyladamantane, 1-Actamido-3,5-dimethyladmantane, N-(3,5-Dimethyladamantan-1-yl)acetamide

A2653-1G A2653-5G
TCIAA2653-1GEA 846 DKK
TCIAA2653-1G TCIAA2653-5G
N-Acetyl-3,5-dimethyl-1-adamantanamine ≥97.0% (by GC)
N-Acetyl-3,5-dimethyl-1-adamantanamine
Formel: C₁₄H₂₃NO
Molekylvægt: 221.34 g/mol
MDL Number: MFCD06656139
CAS-nummer: 19982-07-1

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Melting point 110.0 to 115.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR