Acetamidoxime ≥97.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

A2646-5G A2646-25G
TCIAA2646-5GEA 419 DKK
TCIAA2646-5G TCIAA2646-25G
Acetamidoxime ≥97.0% (by GC, titration analysis)
Acetamidoxime
Formel: C₂H₆N₂O
MDL Number: MFCD00603514
CAS-nummer: 22059-22-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=28f82f1ebb37e2cfde585c37d0e52e4477e0511a&vid=de30d743010361c03a1a36bf6f9bd20d3b76eb79&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR