5-Amino-1-(2-hydroxyethyl)pyrazole ≥99.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 3-Amino-2-pyrazoleethanol

A2619-5G A2619-25G
TCIAA2619-5GEA 247 DKK
TCIAA2619-5G TCIAA2619-25G
5-Amino-1-(2-hydroxyethyl)pyrazole ≥99.0% (by GC)
5-Amino-1-(2-hydroxyethyl)pyrazole
Formel: C₅H₉N₃O
Molekylvægt: 127.15 g/mol
Kogepunkt: 140°C
MDL Number: MFCD08063809
CAS-nummer: 73616-27-0

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 99.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 107.0 to 109.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR