Azilsartan ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-Ethoxy-1-((2'-(5-oxo-2,5-dihydro-1,2,4-oxadiazol-3-yl)-[1,1'-biphenyl]-4-yl)methyl)-1H-benzo[d]imidazole-7-carboxylic acid

A2601-200MG A2601-1G
TCIAA2601-200MGEA 559 DKK
TCIAA2601-200MG TCIAA2601-1G
Azilsartan ≥98.0% (by HPLC, titration analysis)
Azilsartan
Formel: C₂₅H₂₀N₄O₅
Opbevaringstemperatur: Køleskab
MDL Number: MFCD20278186
CAS-nummer: 147403-03-0

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR