trans-4-[4-(4-Pentylcyclohexyl)phenyl]benzonitrile ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
4'-(trans-4-Pentylcyclohexyl)-[1,1'-biphenyl]-4-carbonitrile
A2568-1G
A2568-5G
TCIAA2568-1GEA
411
DKK
TCIAA2568-1G
TCIAA2568-5G
trans-4-[4-(4-Pentylcyclohexyl)phenyl]benzonitrile ≥98.0% (by GC)
trans-4-[4-(4-Pentylcyclohexyl)phenyl]benzonitrile
Formel:
C₂₄H₂₉N Molekylvægt: 331.50 g/mol Kogepunkt: 211°C |
MDL Number:
MFCD01076317 CAS-nummer: 68065-81-6 EINECS: 268-333-6 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...