3-Amino-3-(4-methoxyphenyl)propionic acid ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

A2532-1G A2532-5G
TCIAA2532-1GEA 723 DKK
TCIAA2532-1G TCIAA2532-5G
3-Amino-3-(4-methoxyphenyl)propionic acid ≥98.0% (by HPLC, titration analysis)
3-Amino-3-(4-methoxyphenyl)propionic acid
Formel: C₁₀H₁₃NO₃
Smeltepunkt: 232°C
MDL Number: MFCD00187209
CAS-nummer: 5678-45-5

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR