Aloe emodin ≥97.0% (by HPLC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Aloe-emodin, 3-Hydroxymethylchrysazine, 1,8-Dihydroxy-3-(hydroxymethyl)anthracene-9,10-dione, 1,8-Dihydroxy-3-(hydroxymethyl)anthraquinone
A2491-100MG
A2491-1G
TCIAA2491-100MGEA
386
DKK
TCIAA2491-100MG
TCIAA2491-1G
Aloe emodin ≥97.0% (by HPLC)
Aloe emodin
Formel:
C₁₅H₁₀O₅ Molekylvægt: 270.24 g/mol Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00017373 CAS-nummer: 481-72-1 EINECS: 207-571-7 Merck Index: 14,00306 |
Specifikationer Testresultater
Appearance | Light yellow to Brown powder to lump |
Purity (HPLC) | min. 97.0 area% |
Purity (Neutralization titration) | min. 96.0 % |
Melting point | 220.0 to 224.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...