(Z)-2-(2-Amino-4-thiazolyl)-2-(methoxyimino)acetic acid ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
A2478-5G
A2478-25G
TCIAA2478-5GEA
288
DKK
TCIAA2478-5G
TCIAA2478-25G
(Z)-2-(2-Amino-4-thiazolyl)-2-(methoxyimino)acetic acid ≥98.0% (by HPLC, titration analysis)
(Z)-2-(2-Amino-4-thiazolyl)-2-(methoxyimino)acetic acid
Formel:
C₆H₇N₃O₃S Smeltepunkt: 204°C |
MDL Number:
MFCD00010416 CAS-nummer: 65872-41-5 EINECS: 265-957-0 |
|
Specifikationer Testresultater
Appearance | White to Light yellow powder to crystalline |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...