Acemetacin ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
[1-(4-Chlorobenzoyl)-5-methoxy-2-methylindol-3-yl]acetoxyacetic Acid, 2-(2-(1-(4-Chlorobenzoyl)-5-methoxy-2-methyl-1H-indol-3-yl)acetoxy)acetic acid, 1-(4-Chlorobenzoyl)-5-methoxy-2-methylindole-3-acetic Acid Carboxymethyl Ester
A2452-25G
A2452-5G
A2452-1G
TCIAA2452-25GEA
2210
DKK
TCIAA2452-25G
TCIAA2452-5G
TCIAA2452-1G
Acemetacin ≥98.0% (by GC, titration analysis)
Acemetacin
Formel:
C₂₁H₁₈ClNO₆ Molekylvægt: 415.83 g/mol |
MDL Number:
MFCD00151473 CAS-nummer: 53164-05-9 EINECS: 258-403-4 UN nummer: 2811 ADR: 6.1 II Merck Index: 14,00027 |
Specifikationer Testresultater
Appearance | Light yellow to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 151.0 to 154.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...