Amlexanox ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 2-Amino-7-isopropyl-5-oxo-5H-chromeno[2,3-b]pyridine-3-carboxylic acid, 2-Amino-7-isopropyl-5-oxochromeno[2,3-b]pyridine-3-carboxylic Acid

A2401-1G A2401-5G
TCIAA2401-1GEA 411 DKK
TCIAA2401-1G TCIAA2401-5G
Amlexanox ≥98.0% (by HPLC, titration analysis)
Amlexanox
Formel: C₁₆H₁₄N₂O₄
Smeltepunkt: 300°C
MDL Number: MFCD00864790
CAS-nummer: 68302-57-8
Merck Index: 14,00490

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR