1-β-D-Arabinofuranosyluracil ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

A2356-1G A2356-5G
TCIAA2356-1GEA 132 DKK
TCIAA2356-1G TCIAA2356-5G
1-β-D-Arabinofuranosyluracil ≥98.0% (by HPLC, titration analysis)
1-beta-D-Arabinofuranosyluracil
Formel: C₉H₁₂N₂O₆
Smeltepunkt: 222°C
Opbevaringstemperatur: Fryser
MDL Number: MFCD00065998
CAS-nummer: 3083-77-0
EINECS: 221-386-9

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D +118.0 to +123.0 deg (C=0.5, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR