5-Amino-1,2,3-thiadiazole ≥98.0% (by HPLC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
A2354-5G
A2354-25G
TCIAA2354-5GEA
403
DKK
TCIAA2354-5G
TCIAA2354-25G
5-Amino-1,2,3-thiadiazole ≥98.0% (by HPLC)
5-Amino-1,2,3-thiadiazole
Formel:
C₂H₃N₃S |
MDL Number:
MFCD00082810 CAS-nummer: 4100-41-8 EINECS: 223-868-4 |
Specifikationer Testresultater
Appearance | Light yellow to Brown powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...