(R)-3-Amino-1-N-Cbz-pyrrolidine hydrochloride ≥98.0% (by HPLC, total nitrogen)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
(R)-3-Amino-1-carbobenzoxypyrrolidine hydrochloride, (R)-1-Cbz-3-aminopyrrolidine Hydrochloride, (R)-3-Amino-1-benzyloxycarbonylpyrrolidine hydrochloride, (R)-3-Amino-1-Cbz-pyrrolidine hydrochloride
A2323-1G
A2323-5G
TCIAA2323-1GEA
403
DKK
TCIAA2323-1G
TCIAA2323-5G
(R)-3-Amino-1-N-Cbz-pyrrolidine hydrochloride ≥98.0% (by HPLC, total nitrogen)
(R)-3-Amino-1-N-Cbz-pyrrolidine hydrochloride
Formel:
C₁₂H₁₇ClN₂O₂ |
MDL Number:
MFCD09038802 CAS-nummer: 870621-17-3 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Optical purity (LC) | min. 96.0 ee% |
Purity (with Total Nitrogen) | min. 98.0 % |
Specific rotation [a]20/D | -13.0 to -16.0 deg (C=1, H2O) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...