(S)-3-(4-Aminophenyl)-2-((tert-butoxycarbonyl)amino)propanoic acid ≥96.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

A2131-1G A2131-5G
TCIAA2131-1GEA 279 DKK
TCIAA2131-1G TCIAA2131-5G
(S)-3-(4-Aminophenyl)-2-((tert-butoxycarbonyl)amino)propanoic acid ≥96.0% (by HPLC, titration analysis)
(S)-3-(4-Aminophenyl)-2-((tert-butoxycarbonyl)amino)propanoic acid
Formel: C₁₄H₂₀N₂O₄
MDL Number: MFCD00038139
CAS-nummer: 55533-24-9

Bestil nu

Specifikationer Testresultater

Purity (HPLC) min. 96.0 area%
Purity (Nonaqueous Titration) min. 96.0 %
Specific rotation [a]20/D +23.0 to +29.0 deg (C=1, MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR