2-Amino-2'-nitrodiphenyl sulphide ≥98.0% (by HPLC, total nitrogen)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
2-Aminophenyl 2-Nitrophenyl sulphide, 2-(2-Nitrophenylthio)aniline
A1985-25G
A1985-5G
TCIAA1985-25GEA
1330
DKK
TCIAA1985-25G
TCIAA1985-5G
2-Amino-2'-nitrodiphenyl sulphide ≥98.0% (by HPLC, total nitrogen)
2-Amino-2'-nitrodiphenyl sulphide
Formel:
C₁₂H₁₀N₂O₂S Molekylvægt: 246.28 g/mol |
MDL Number:
MFCD00121816 CAS-nummer: 19284-81-2 |
Specifikationer Testresultater
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 84.0 to 88.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...