4-Amino-5-chloro-2-ethoxybenzoic acid ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
4-Amino-5-chloro-2-ethoxybenzoic Acid
A1941-25G
A1941-5G
TCIAA1941-25GEA
329
DKK
TCIAA1941-25G
TCIAA1941-5G
4-Amino-5-chloro-2-ethoxybenzoic acid ≥98.0% (by HPLC, titration analysis)
4-Amino-5-chloro-2-ethoxybenzoic acid
Formel:
C₉H₁₀ClNO₃ |
MDL Number:
MFCD03840527 CAS-nummer: 108282-38-8 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 167.0 to 171.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...