Trimethylsilylmethyl acetate ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
(Trimethylsilyl)methyl acetate, Acetic Acid Trimethylsilylmethyl Ester
A1613-25G
A1613-5G
TCIAA1613-25GEA
903
DKK
TCIAA1613-25G
TCIAA1613-5G
Trimethylsilylmethyl acetate ≥98.0% (by GC)
Trimethylsilylmethyl acetate
Formel:
C₆H₁₄O₂Si Molekylvægt: 146.26 g/mol Kogepunkt: 137°C Flammepunkt: 20°C |
MDL Number:
MFCD00009650 CAS-nummer: 2917-65-9 EINECS: 220-849-2 UN nummer: 3272 ADR: 3 II |
Specifikationer Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...