Diethoxymethyl acetate ≥97.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

A1608-25G A1608-5G
TCIAA1608-25GEA 1430 DKK
TCIAA1608-25G TCIAA1608-5G
Diethoxymethyl acetate ≥97.0% (by GC)
Diethoxymethyl acetate
Formel: C₇H₁₄O₄
Flammepunkt: 27°C
MDL Number: MFCD00009229
CAS-nummer: 14036-06-7
EINECS: 237-873-4
UN nummer: 3272
ADR: 3,III

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR