(R)-4-Amino-2-methyl-1-butanol ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

A1576-1G A1576-5G
TCIAA1576-1GEA 517 DKK
TCIAA1576-1G TCIAA1576-5G
(R)-4-Amino-2-methyl-1-butanol ≥98.0% (by GC, titration analysis)
(R)-4-Amino-2-methyl-1-butanol
Formel: C₅H₁₃NO
Kogepunkt: 80°C
MDL Number: MFCD03093614
CAS-nummer: 88390-32-3

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D +14.0 to +15.0 deg (neat)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR