2-Amino-5-bromothiazole hydrobromide 95,0 Titrimetric

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-Amino-5-bromothiazole monohydrobromide, 2-Amino-5-bromothiazole HBr

A1573-25G A1573-5G
TCIAA1573-25GEA 747 DKK
TCIAA1573-25G TCIAA1573-5G
2-Amino-5-bromothiazole hydrobromide 95,0 Titrimetric
2-Amino-5-bromothiazole hydrobromide
Formel: C₃H₃BrN₂S
Molekylvægt: 259.95 g/mol
MDL Number: MFCD00012712
CAS-nummer: 61296-22-8
EINECS: 262-699-0

Bestil nu

Specifikationer Testresultater

Appearance White to Gray to Brown powder to crystal
Purity (Nonaqueous Titration) min. 95.0 %
Melting point 157.0 to 163.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR