3-Acetoxy-2-methylbenzoic acid ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 3-Acetoxy-o-toluic acid, 3-Acetoxy-o-toluic Acid, 3-Acetoxy-2-methylbenzoic Acid

A1558-25G A1558-5G
TCIAA1558-25GEA 764 DKK
TCIAA1558-25G TCIAA1558-5G
3-Acetoxy-2-methylbenzoic acid ≥98.0% (by HPLC, titration analysis)
3-Acetoxy-2-methylbenzoic acid
Formel: C₁₀H₁₀O₄
Molekylvægt: 194.19 g/mol
MDL Number: MFCD00957176
CAS-nummer: 168899-58-9

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 148.0 to 152.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR