4-Vinylphenyl acetate ≥98.0% (by GC) stabilised
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
Acetic Acid 4-Vinylphenyl Ester, 4-Acetoxystyrene
Stabilization:
Stabilised with 4-tert-butylpyrocatechol (TBC)
A1551-25G
A1551-5G
TCIAA1551-25GEA
714
DKK
TCIAA1551-25G
TCIAA1551-5G
4-Vinylphenyl acetate ≥98.0% (by GC) stabilised
4-Vinylphenyl acetate
Formel:
C₁₀H₁₀O₂ Molekylvægt: 162.19 g/mol Flammepunkt: 87°C |
MDL Number:
MFCD00075734 CAS-nummer: 2628-16-2 |
Specifikationer Testresultater
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...