2-Ethylphenyl acetate ≥97.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

A1549-25G A1549-5G
TCIAA1549-25GEA 1280 DKK
TCIAA1549-25G TCIAA1549-5G
2-Ethylphenyl acetate ≥97.0% (by GC)
2-Ethylphenyl acetate
Formel: C₁₀H₁₂O₂
MDL Number: MFCD02093429
CAS-nummer: 3056-59-5

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR