Allylpalladium(II) chloride dimer ≥97.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Di-mu-chlorobis[(eta-allyl)palladium(II)], Allylpalladium(II) chloride, Allylpalladium chloride dimer

A1479-1G A1479-500MG
TCIAA1479-1GEA 2340 DKK
TCIAA1479-1G TCIAA1479-500MG
Allylpalladium(II) chloride dimer ≥97.0% (by titrimetric analysis)
Allylpalladium(II) chloride dimer
Formel: C₆H₁₀Cl₂Pd₂
MDL Number: MFCD00044874
CAS-nummer: 12012-95-2
EINECS: 234-579-8

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (Argentmetric Titration) min. 97.0 %
Purity (Chelometric Titration) min. 97.0 %
Solubility in THF almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR