(S)-(+)-O-Acetylmandelic acid ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: (S)-2-Acetoxy-2-phenylacetic acid, (S)-(+)-α-Acetoxyphenylacetic Acid, (+)-O-Acetyl-L-mandelic Acid

A1454-25G A1454-5G
TCIAA1454-25GEA 1210 DKK
TCIAA1454-25G TCIAA1454-5G
(S)-(+)-O-Acetylmandelic acid ≥98.0% (by titrimetric analysis)
(S)-(+)-O-Acetylmandelic acid
Formel: C₁₀H₁₀O₄
Molekylvægt: 194.19 g/mol
MDL Number: MFCD00064215
CAS-nummer: 7322-88-5

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 96.0 to 102.0 deg-C
Specific rotation [a]20/D +151 to +155 deg (C=2, Aceton)
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR