(S)-(+)-2-Amino-4-bromobutyric acid hydrobromide ≥98.0% (by total nitrogen and titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: (S)-2-Amino-4-bromobutanoic acid hydrobromide

A1450.0005 A1450-1G A1450-5G
TCIAA1450.0005EA 3870 DKK
TCIAA1450.0005 TCIAA1450-1G TCIAA1450-5G
(S)-(+)-2-Amino-4-bromobutyric acid hydrobromide ≥98.0% (by total nitrogen and titration analysis)
(S)-(+)-2-Amino-4-bromobutyric acid hydrobromide
Formel: C₄H₈BrNO₂
Smeltepunkt: 184°C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD01631290
CAS-nummer: 15159-65-6

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Specific rotation [a]20/D +14.0 to +19.0 deg (C=1, MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR