Sodium anthraquinone-1-sulphonate ≥98.0% (by HPLC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Anthraquinone-1-sulphonic acid Sodium Salt, 9,10-Dihydro-9,10-dioxo-1-anthraquinonesulphonic acid sodium salt, 1-Anthraquinonesulphonic acid sodium salt, Sodium 9,10-dioxo-9,10-dihydroanthracene-1-sulphonate

A1428-25G A1428-250G
TCIAA1428-25GEA 165 DKK
TCIAA1428-25G TCIAA1428-250G
Sodium anthraquinone-1-sulphonate ≥98.0% (by HPLC)
Sodium anthraquinone-1-sulphonate
Formel: C₁₄H₈O₅S
Molekylvægt: 310.25 g/mol
MDL Number: MFCD00037145
CAS-nummer: 128-56-3
EINECS: 204-894-5

Bestil nu

Specifikationer Testresultater

Appearance Light orange to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Solubility in hot Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR