N-(tert-Butoxycarbonyl)-1,5-diaminopentane ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: N-Boc-1,5-diaminopentane, tert-Butyl N-(5-Aminopentyl)carbamate, tert-Butyl (5-aminopentyl)carbamate, N-(5-Aminoamyl)carbamic Acid tert-Butyl Ester, N-(tert-Butoxycarbonyl)-1,5-pentanediamine, N-(5-Aminopentyl)carbamic Acid tert-Butyl Ester, NH2-C5-NH-Boc, N-Boc-1,5-pentanediamine

A1374-1G A1374-5G
TCIAA1374-1GEA 624 DKK
TCIAA1374-1G TCIAA1374-5G
N-(tert-Butoxycarbonyl)-1,5-diaminopentane ≥98.0% (by titrimetric analysis)
N-(tert-Butoxycarbonyl)-1,5-diaminopentane
Formel: C₁₀H₂₂N₂O₂
MDL Number: MFCD00210020
CAS-nummer: 51644-96-3

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 96.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR