N-Boc-1,3-propanediamine ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: N-(3-Aminopropyl)carbamic Acid tert-Butyl Ester, tert-Butyl N-(3-Aminopropyl)carbamate, N-Boc-1,3-propanediamine, tert-Butyl (3-aminopropyl)carbamate, N-1-Boc-1,3-diaminopropane·HCl, N-(tert-Butoxycarbonyl)-1,3-propanediamine, N-Boc-1,3-diaminopropane

A1372-1G A1372-5G
TCIAA1372-1GEA 263 DKK
TCIAA1372-1G TCIAA1372-5G
N-Boc-1,3-propanediamine ≥98.0% (by GC)
N-Boc-1,3-propanediamine
Formel: C₈H₁₈N₂O₂
MDL Number: MFCD00210021
CAS-nummer: 75178-96-0
UN nummer: 3259
ADR: 8,III

Bestil nu

Specifikationer Testresultater

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR