(S)-(-)-2-Acetoxypropionyl chloride ≥97.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
A1297-1G
A1297-5G
TCIAA1297-1GEA
304
DKK
TCIAA1297-1G
TCIAA1297-5G
(S)-(-)-2-Acetoxypropionyl chloride ≥97.0% (by GC, titration analysis)
(S)-(-)-2-Acetoxypropionyl chloride
Formel:
C₅H₇ClO₃ Kogepunkt: 78°C Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00145252 CAS-nummer: 36394-75-9 EINECS: 420-610-4 UN nummer: 3265 ADR: 8,II |
Specifikationer Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 97.0 % |
Optical purity (GC) | min. 95.0 ee% |
Purity (Argentmetric Titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...