N-(3-Aminopropyl)diethanolamine ≥90.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: N,N-Bis(2-hydroxyethyl)-1,3-diaminopropane

A1280-25G A1280-500G
TCIAA1280-25GEA 526 DKK
TCIAA1280-25G TCIAA1280-500G
N-(3-Aminopropyl)diethanolamine ≥90.0% (by GC)
N-(3-Aminopropyl)diethanolamine
Formel: C₇H₁₈N₂O₂
Kogepunkt: 207°C
Flammepunkt: 138°C
MDL Number: MFCD00047971
CAS-nummer: 4985-85-7
EINECS: 225-642-0
UN nummer: 2735
ADR: 8,III

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 90.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR