Hexyl acrylate ≥96.0% (by GC) stabilised

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel


Stabilization: stabiliseret med hydroquinon

A1257-25ML A1257-500ML
TCIAA1257-25MLEA 247 DKK
TCIAA1257-25ML TCIAA1257-500ML
Hexyl acrylate ≥96.0% (by GC) stabilised
Hexyl acrylate
Formel: C₉H₁₆O₂
Kogepunkt: 89°C
Flammepunkt: 68°C
MDL Number: MFCD00048881
CAS-nummer: 2499-95-8
EINECS: 219-698-5
UN nummer: 3082
ADR: 9,III

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR