(3R)-(-)-1-Benzyl-3-aminopyrrolidine ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: (R)-1-Benzylpyrrolidin-3-amine, (R)-(-)-1-Benzyl-3-aminopyrrolidine

A1173-10G A1173-25G
TCIAA1173-10GEA 805 DKK
TCIAA1173-10G TCIAA1173-25G
(3R)-(-)-1-Benzyl-3-aminopyrrolidine ≥98.0% (by GC, titration analysis)
(3R)-(-)-1-Benzyl-3-aminopyrrolidine
Formel: C₁₁H₁₆N₂
Molekylvægt: 176.26 g/mol
Flammepunkt: 140°C
MDL Number: MFCD00082638
CAS-nummer: 114715-39-8
UN nummer: 2735
ADR: 8 III

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Optical purity (GC) min. 98.0 ee%
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D -9.3 to -9.6 deg (C=5, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR