Aminomalononitrile-p-toluenesulphonate ≥98.0% (by titrimetric analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
Aminomalononitrile p-Toluenesulphonate, Aminomalononitrile p-Toluenesulphonic acid, Aminomalononitrile p-Toluenesulfonic acid, Dicyanomethylammonium p-Toluenesulphonate, 2-Aminomalononitrile 4-methylbenzenesulphonate, Dicyanomethylammonium p-Toluenesulfonate
A1119-25G
A1119-5G
TCIAA1119-25GEA
1880
DKK
TCIAA1119-25G
TCIAA1119-5G
Aminomalononitrile-p-toluenesulphonate ≥98.0% (by titrimetric analysis)
Aminomalononitrile-p-toluenesulphonate
Formel:
C₃H₃N₃ Molekylvægt: 253.28 g/mol Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00012848 CAS-nummer: 5098-14-6 EINECS: 225-817-1 UN nummer: 2811 ADR: 6.1 III |
Specifikationer Testresultater
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (Neutralization titration) | min. 98.0 % |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...