2-Amino-6-chloropyrimidin-4(3H)-one ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-Amino-4-chloro-6-pyrimidinol, 2-Amino-4-chloro-6-hydroxypyrimidine

A1109-25G A1109-5G
TCIAA1109-25GEA 1970 DKK
TCIAA1109-25G TCIAA1109-5G
2-Amino-6-chloropyrimidin-4(3H)-one ≥98.0% (by HPLC, titration analysis)
2-Amino-6-chloropyrimidin-4(3H)-one
Formel: C₄H₄ClN₃O
MDL Number: MFCD00006093
CAS-nummer: 1194-21-4
EINECS: 214-785-4

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR