3-Aminopyrrolidine ≥99.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

A1104-25G A1104-5G
TCIAA1104-25GEA 2380 DKK
TCIAA1104-25G TCIAA1104-5G
3-Aminopyrrolidine ≥99.0% (by GC, titration analysis)
3-Aminopyrrolidine
Formel: C₄H₁₀N₂
Molekylvægt: 86.14 g/mol
MDL Number: MFCD00059018
CAS-nummer: 79286-79-6
UN nummer: 2735
ADR: 8 III

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %
Purity (Nonaqueous Titration) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR