2-Methylpentyl acetate ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Acetic Acid 2-Methylpentyl Ester
A1079-25ML
A1079-500ML
TCIAA1079-25MLEA
222
DKK
TCIAA1079-25ML
TCIAA1079-500ML
2-Methylpentyl acetate ≥98.0% (by GC)
2-Methylpentyl acetate
Formel:
C₈H₁₆O₂ Kogepunkt: 160°C |
MDL Number:
MFCD00060103 CAS-nummer: 7789-99-3 UN nummer: 3272 ADR: 3,III |
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...