4-Methoxybenzyl acetate ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
A0886-25ML
A0886-500ML
TCIAA0886-25MLEA
132
DKK
TCIAA0886-25ML
TCIAA0886-500ML
4-Methoxybenzyl acetate ≥98.0% (by GC)
4-Methoxybenzyl acetate
Formel:
C₁₀H₁₂O₃ Kogepunkt: 137°C Flammepunkt: 140°C |
MDL Number:
MFCD00038509 CAS-nummer: 104-21-2 EINECS: 203-185-8 |
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...