O-Acetylricinoleic acid methyl ester ≥80.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
A0879-25ML
A0879-500ML
TCIAA0879-25MLEA
132
DKK
TCIAA0879-25ML
TCIAA0879-500ML
O-Acetylricinoleic acid methyl ester ≥80.0% (by GC)
O-Acetylricinoleic acid methyl ester
Formel:
C₂₁H₃₈O₄ Kogepunkt: 260°C Flammepunkt: 198°C |
MDL Number:
MFCD00026204 CAS-nummer: 140-03-4 EINECS: 205-392-9 |
Specifikationer Testresultater
Appearance | Colorless to Orange to Green clear liquid |
Purity (GC) | min. 80.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...