3-Methoxybutyl acetate ≥99.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
Acetic Acid 3-Methoxybutyl Ester
A0759.0500
A0759-25ML
A0759-500ML
TCIAA0759.0500EA
733
DKK
TCIAA0759.0500
TCIAA0759-25ML
TCIAA0759-500ML
3-Methoxybutyl acetate ≥99.0% (by GC)
3-Methoxybutyl acetate
Formel:
C₇H₁₄O₃ Molekylvægt: 146.19 g/mol Kogepunkt: 172°C Flammepunkt: 62°C |
MDL Number:
MFCD00043928 CAS-nummer: 4435-53-4 EINECS: 224-644-9 |
Specifikationer Testresultater
Appearance | Colorless clear liquid |
Purity (GC) | min. 99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...